Få mere at vide
Invitrogen™ APOOL Polyclonal Antibody
Beskrivelse
Antibody detects endogenous levels of total ApoOL.
This gene encodes a protein which contains an apolipoprotein O superfamily domain. This domain is found on proteins in circulating lipoprotein complexes. [provided by RefSeq, Sep 2011].
Tekniske data
Tekniske data
| Antigen | APOOL |
| Applikationer | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugeret | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | APOOL |
| Genadgangsnr. | Q6UXV4, Q78IK4 |
| Gene Alias | 6720473G16Rik; 9430083G14Rik; AAIR8193; apolipoprotein O like; apolipoprotein O-like; Apool; CXorf33; E130106L15Rik; FAM121A; family with sequence similarity 121A; MIC27; MICOS complex subunit MIC27; Protein FAM121A; RGD1549725; UNQ8193; UNQ8193/PRO23204 |
| Gensymboler | APOOL |
| Vis mere |