missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Apolipoprotein M Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112362
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Apolipoprotein M Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized.
Tekniske data
| Apolipoprotein M | |
| Polyclonal | |
| Unconjugated | |
| APOM | |
| 1190010O19Rik; alternative name: G3a, NG20; Apo M; Apolipoprotein M; APOM; apo-M; G3A; HSPC336; MGC22400; NG20; NG20 like protein; NG20-like protein; protein G3a; Protein Px | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55937, 55938 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O95445, Q9Z1R3 | |
| APOM | |
| Recombinant Human Apolipoprotein M protein (23-188AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion