missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Apolipoprotein B/ApoB Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-7616
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Apolipoprotein B/ApoB Polyclonal specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Sandwich ELISA.
Tekniske data
| Apolipoprotein B/ApoB | |
| Polyclonal | |
| Unconjugated | |
| 0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 with 0.01% Sodium Azide | |
| Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
| Goat | |
| 92.33 kDa | |
| 0.2 mg | |
| Lipid and Metabolism | |
| 338 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1:200-1:1000, ELISA 1:2000-1:10000, Immunohistochemistry 1:50-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin, Sandwich ELISA 1:100-1:2000 | |
| Q7Z7Q0 | |
| APOB | |
| Apolipoprotein B/ApoB was isolated from human plasma by density gradient centrifugation followed by HPLC purification. (Uniprot: Q7Z7Q0) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Typically less than 1% cross reactivity against other types of apoLipoprotein was detected by ELISA against purified standards. This antibody reacts with human apoLipoprotein B and has negligible cross-reactivity with Type A-I, A-II, C-I, C-II, C-III, E and J apoLipoproteins. Specific cross reaction of anti-apoLipoprotein antibodies with antigens from other species has not been determined. Non-specific cross reaction of anti-apoLipoprotein antibodies with other human serum proteins is negligible. | |
| Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction