missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Apolipoprotein A-II/ApoA2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-1303
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Apolipoprotein A-II/ApoA2 Polyclonal specifically detects Apolipoprotein A-II/ApoA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Tekniske data
| Apolipoprotein A-II/ApoA2 | |
| Polyclonal | |
| Unconjugated | |
| 0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 with 0.01% Sodium Azide | |
| APOA2, apoAII, Apo-AII, ApoA-II, Apolipoprotein A2, apolipoprotein A-II | |
| Goat | |
| 11.17 kDa | |
| 0.1 mg | |
| Primary | |
| Typically less than 1% cross reactivity against other types of apolipoprotein was detected by ELISA against purified standards. This antibody reacts with human apolipoprotein A-II and has negligible cross-reactivity with Type A-I, B, C-I, C-II, C-III, E and J apolipoproteins. Specific cross reaction of anti-apolipoprotein antibodies with antigens from other species has not been determined. Non-specific cross reaction of anti-apolipoprotein antibodies with other human serum proteins is negligible. | |
| Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1:5000-1:10000, ELISA 1:5000-1:10000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500 | |
| P02652, P02652, P02652, P02652 | |
| APOA2 | |
| Apolipoprotein A-II/ApoA2 was isolated from human plasma by density gradient centrifugation followed by HPLC purification. (Uniprot: P02652) | |
| Affinity Purified | |
| RUO | |
| 336 | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion