Få mere at vide
β-Apo-oxytetracycline (>80%), TRC
Artikelnummer. 30334669
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30334669 10 mg
30340657 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30334669 Leverandør Toronto Research Chemicals Leverandørnr. TRCA72903810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Beta-apo-oxytetracycline
CAS 18751-99-0
Molekylær formel C22 H22 N2 O8
Mængde 10 mg
Synonym Beta-apo-oxytetracycline, 1-Cyclohexene-1-carboxamide, 4-[(3R)-1,3-dihydro-4,5-dihydroxy-9-methyl-3-oxonaphtho[2,3-c]furan-1-yl]-3-(dimethylamino)-2,5-dihydroxy-6-oxo-, (3S,4S,5R)-, (3S,4S,5R)-4-[(1R)-1,3-Dihydro-4,5-dihydroxy-9-methyl-3-oxonaphtho[2,3-c]furan-1-yl]-3-(dimethylamino)-2,5-dihydroxy-6-oxo-1-cyclohexene-1-carboxamide, β-Apo-5-oxytetracycline, β-Apooxytetracycline, β-Apoterramycin
InChI formel InChI=1S/C22H22N2O8/c1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)20(32-22(12)31)13-15(24(2)3)17(27)14(21(23)30)19(29)18(13)28/h4-6,13,15,18,20,25-28H,1-3H3,(H2,23,30)/t13-,15-,18+,20-/m0/s1
SMIL CN(C)[C@H]1[C@@H]([C@@H](O)C(=O)C(=C1O)C(=O)N)[C@H]2OC(=O)c3c(O)c4c(O)cccc4c(C)c23
IUPAC navn (3S,4S,5R)-4-[(1R)-4,5-dihydroxy-9-methyl-3-oxo-1H-benzo[f][2]benzofuran-1-yl]-3-(dimethylamino)-2,5-dihydroxy-6-oxocyclohexene-1-carboxamide
Molekylvægt (g/mol) 442.42
Formel vægt 442.1376