missing translation for 'onlineSavingsMsg'
Learn More
APNEA, MedChemExpress Product Code.: 16427588

APNEA, MedChemExpress

Product Code. p-200101313
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16427588

Brand: Medchem Express HY18687

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress APNEA (N6-[2-(4-Aminophenyl)ethyl]adenosine) is a potent, non-selective A3 adenosine receptor agonist.

APNEA (N6-[2-(4-Aminophenyl)ethyl]adenosine) is a potent, non-selective A3 adenosine receptor agonist.

Specifications

Chemical Name or Material APNEA
CAS 89705-21-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H22N6O4
Quantity 10 mg
Synonym N6-[2-(4-Aminophenyl)ethyl]adenosine
Solubility Information DMSO : 250 mg/mL (646.98 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H]([C@H]([C@H](N2C=NC3=C2N=CN=C3NCCC4=CC=C(N)C=C4)O1)O)O
Molecular Weight (g/mol) 386.41
Formula Weight 386.41
Percent Purity 95.01%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
APNEA, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.