Få mere at vide
Apalutamide-d3, TRC
Artikelnummer. 30336855
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30336855 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336855 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7261231MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Apalutamide-d3
CAS 1638885-61-6
Molekylær formel C21 D3 H12 F4 N5 O2 S
Mængde 1 mg
Synonym 4-[7-[6-Cyano-5-(trifluoromethyl)-3-pyridinyl]-8-oxo-6-thioxo-5,7-diazaspiro[3.4]oct-5-yl]-2-fluoro-N-(methyl-d3)-benzamide
InChI formel InChI=1S/C21H15F4N5O2S/c1-27-17(31)13-4-3-11(8-15(13)22)30-19(33)29(18(32)20(30)5-2-6-20)12-7-14(21(23,24)25)16(9-26)28-10-12/h3-4,7-8,10H,2,5-6H2,1H3,(H,27,31)/i1D3
SMIL [2H]C([2H])([2H])NC(=O)c1ccc(cc1F)N2C(=S)N(C(=O)C23CCC3)c4cnc(C#N)c(c4)C(F)(F)F
IUPAC navn 4-[7-[6-cyano-5-(trifluoromethyl)pyridin-3-yl]-8-oxo-6-sulfanylidene-5,7-diazaspiro[3.4]octan-5-yl]-2-fluoro-N-(trideuteriomethyl)benzamide
Molekylvægt (g/mol) 480.453
Formel vægt 480.107