Få mere at vide
Grant Instruments™ ap360 Air Purifier
Artikelnummer. 17200364
Skift visning
Luftstrømshastighed:
360 m3/t.
Artikelnummer. Luftstrømshastighed
17200364 360 m3/t.
1 options
Artikelnummer. 17200364 Leverandør Grant Instruments™ Leverandørnr. AP360
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Grant Instruments' core engineering strengths and manufacturing expertise, together with proven science around ultraviolet light (UVC) as a disinfectant, mean our quiet, high-performance ap360 Air Purifier device is an ideal air hygiene solution for clinical settings and businesses
  • Grant Instruments ap360 Air Purifier is designed to help you keep your staff, patients, and customers safe from airborne infection as well as to remove aerosols, pollen, dust and other particulates
  • UVC lamps provide powerful UV air sterilisation that disrupts the DNA and RNA in bacteria and viruses
  • Large air processing capacity of 360m3/hour enables minimal downtime
  • Fast fan speed with three modes and auto settings
  • Digital timer 1 min to 24 hrs/non-stop can be set to the desired minimum or maximum use
  • Full user protection from direct UV light
  • Compact design to minimise use of space and maximise portability
  • Low power usage for a cost-effective and eco-friendly air purification solution
TRUSTED_SUSTAINABILITY
Tekniske data
Effektivitet 0.9995
Filtertype HEPA13
Luftstrømshastighed 360 m3/t.
Vægt (metrisk) 13.4 Kg
Højde (metrisk) 1050 mm
Dybde (metrisk) 350 mm
Bredde (metrisk) 350 mm
Produkttype Air Purifier
Spænding 120/230 V
Frekvens 50/60 Hz
Certificeringer/compliance EN1822 Standard HEPA13