missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP1G2 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05786JF549
Additional Details : Weight : 0.00970kg
Description
AP1G2 Polyclonal specifically detects AP1G2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
AP1G2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
adaptor-related protein complex 1, gamma 2 subunit, AP-1 complex subunit gamma-like 2, clathrin-associated/assembly/adaptor protein, large, gamma-2, G2AD, Gamma2-adaptin | |
Produced in rabbits immunized with E. coli-derived Human AP1G2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
8906 | |
Store at 4°C in the dark. | |
IgG |