Få mere at vide
Antipyrine-d3 (N-methyl-d3), CDN
Artikelnummer. 30321654
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321654 0.1 g 0.1g
30314576 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321654 Leverandør CDN Leverandørnr. CDND39650.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Antipyrine-d3
CAS 65566-62-3
Molekylær formel C11 2H3 H9 N2 O
Mængde 0.1 g
InChI formel InChI=1 S/C11H12N2O/c1-9-8-11(14)13(12(9)2)10-6-4-3-5-7-10/h3-8 H,1-2H3/i2D3
SMIL [2 H]C([2 H])([2 H])N1N(C(=O)C=C1C)c2ccccc2
IUPAC navn 5-methyl-2-phenyl-1-(trideuteriomethyl)pyrazol-3-one
Molekylvægt (g/mol) 191.24
Formel vægt 191.1138 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity