missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony trichloride R1 solution EU Pharmacopoeia Spectracer, Fisher Chemical™
Click to view available options
Quantity:
100 mL, 10 mL
Packaging:
Glass Bottle
Specifications
Packaging | Glass Bottle |
Quantity | 100 mL, 10 mL |
Grade | EUR Ph Ref: 1007702 |
Chemical Name or Material | Antimony trichloride Solution R1 (Solution I and II) |
Safety and Handling
Product Identifier
- Antimony trichloride Solution R1 (Solution I) European Pharmacopoeia Ref: 1007702. (RP 025)
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Carcinogenicity Category 1B
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'