Learn More
Antimony, plasma standard solution, Specpure™ Sb 1000μg/mL Product Code.: 11308828
Antimony, plasma standard solution, Specpure™ Sb 1000μg/mL
Product Code. p-7082667
50 mL, 50mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Product Code. 11308828
Brand: Thermo Scientific Alfa Aesar 041682.AD
Request Bulk or Custom Format
Sb in H2O/tartaric acid/tr. HNO3
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
Molecular Formula Sb in H2O/tartaric acid/tr. HNO3
MDL Number MFCD00134030
Specifications
Physical Form Liquid
Concentration Sb 1000 ÎĽg/mL
Quantity 50 mL
Chemical Name or Material Antimony, plasma standard solution, Specpure™
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number1546667 Certificate TypeCertificate of Analysis Date07/04/2025 Product Code041682.AE,041682.AD,041682.AP
Lot Number1525086 Certificate TypeCertificate of Analysis Date13/03/2025 Product Code041682.AE,041682.AD,041682.AP
Lot Number1525085 Certificate TypeCertificate of Analysis Date13/03/2025 Product Code041682.AE,041682.AD,041682.AP
Lot Number1521712 Certificate TypeCertificate of Analysis Date15/01/2025 Product Code041682.AE,041682.AD,041682.AP
Lot Number1510708 Certificate TypeCertificate of Analysis Date06/12/2024 Product Code041682.AE,041682.AD,041682.AP
RUO – Research Use Only
Product Content Correction
Product Title
Antimony, plasma standard solution, Specpure™ Sb 1000μg/mL >