missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
Plastic bottle
Description
Antimony(III) sulfide is manufactured for use in safety matches, military ammunition, explosives and fireworks. Also it is used in the production of ruby-colored glass and in plastics as a flame retardant. It is a semiconductor with a direct band gap of 1.8-2.5 eV. With suitable doping p and n type materials can be produced.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 550°C |
Packaging | Glass bottle |
Quantity | 2 g |
UN Number | UN1549 |
Formula Weight | 339.68 |
Percent Purity | 99.999% |
Physical Form | Pieces |
Chemical Name or Material | Antimony(III) sulfide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'