missing translation for 'onlineSavingsMsg'
Få mere at vide

Antimony(III) Oxide, For Analysis, EMSURE™, MilliporeSigma™

Artikelnummer. 15865060
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 kg
Emballage:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Emballage
15865060 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15865060 Leverandør Supelco Leverandørnr. 1078361000
 Begrænset vare
Forventet forsendelse: 18-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
For analysis
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1309-64-4
Molekylær formel O3Sb2
Molekylvægt (g/mol) 291.52
MDL nummer MFCD00011214
InChI nøgle GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym di-Antimony trioxide
IUPAC navn diantimony(3+) trioxidandiide
SMIL [O--].[O--].[O--].[Sb+3].[Sb+3]

Tekniske data

Kemisk navn eller materiale Antimony(III) oxide
Fysisk form Solid
Mængde 1 kg
Formel vægt 291.49
Lugt Odorless
Emballage Plastic bottle
Udseende White solid
Damptryk 13.3 hPa at 660°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.