Få mere at vide
Macherey Nagel Antibiotic Resistance Test Disks, MN 827 ATD
Used to test resistance of pathogens to antibiotics.
Brand:  Macherey Nagel 484001
Produktkode 10338531
  • 271.00 DKK / Pakke med 1000
Kun null tilbage
Forventet forsendelse: 09-02-2026
Tilføj til indkøbskurv
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
  • These products are used in testing the resistance of pathogens to antibiotics
  • For this test the filter paper sections can be impregnated with the antibiotic to be tested and placed on the inoculated nutrient medium
  • Depending on the effectiveness, a smaller or larger zone of inhibition is formed
Tekniske data
Tekniske data
Basis weight: 270 g/m2, Filtration speed: 12 sec., Thickness: 0.7 mm
1000/Pk.
9 mm