missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSV00006W
Additional Details : Weight : 0.01000kg
Description
OSV00006W is specific for VGLUT1 (both isoforms 1 and 2). Reconstitute with 100 μL of sterile water.
a Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.Specifications
VGluT1 | |
Polyclonal | |
SLC17A7 | |
Vesicular glutamate transporter 1, VGlU1, Solute carrier family 17 member 7, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, Bnpi | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9P2U7, Q3TXX4, Q62634 | |
SLC17A7 | |
A synthetic peptide from aa region 500-560 of rat VGluT1 conjugated to an immunogenic carrier protein was used as the antigen | |
RUO | |
116638, 57030, 72961 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |