missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
anti-VAPA, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand: Abnova H00009218-PW1.1Set
Indeholder: Antibody pair for IP: rabbit polyclonal anti-VAPA (300μL), Antibody pair for WB: mouse purified polyclonal anti-VAPA (50μg)
Tekniske data
| VAPA | |
| Monoclonal/Polyclonal | |
| This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
| MGC3745/VAP-33/VAP-A/VAP33/hVAP-33 | |
| Mouse, Rabbit | |
| RUO | |
| 9218 | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| VAPA | |
| VAPA | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-VAPA (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-VAPA (50 ug) | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion