missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V5 Tag Monoclonal Antibody (E10/V4RR), DyLight™ 488, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515253D488
Additional Details : Weight : 0.01000kg
Description
MA5-15253-D488 has been successfully used for immunofluorescence. DyLight 488 has an excitation/emission of 493/518 nm.
V5 Tag Monoclonal antibody specifically detects V5 Tag in Tag samples. It is validated for Immunocytochemistry.Specifications
V5 Tag | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.02% sodium azide | |
Mouse | |
Protein A | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Immunocytochemistry | |
E10/V4RR | |
DyLight 488 | |
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
GKPIPNPLLGLDST (V5) synthetic peptide conjugated to KLH | |
50 μL | |
Primary | |
Antibody | |
IgG1 |