missing translation for 'onlineSavingsMsg'
Learn More

TurboGFP Rabbit anti-Tag, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA522688

Additional Details : Weight : 1.24740kg

Product Code. 13299698

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

PA5-22688 can be used to recognize denatured and non-denatured TurboGFP and CopGFP in immunocytochemistry, Western blot, and ELISA assays. TurboGFP is an improved variant of the green fluorescent protein CopGFP cloned from copepod Pontellina plumata (Arthropoda; Crustacea; Maxillopoda; Copepoda) Reconstitute with sterile water or 50% glycerol to a concentration of 1 mg/mL. Store at -20 C for one year; reconstituted with sterile water, antibody can be stored at +2 - +8°C for three months; reconstituted with 50% glycerol, antibody can be stored at -20°C for six months.

The jellyfish Aequorea victoria contains green fluorescent protein (GFP) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organinsms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcelluclar compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used etensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
Specifications

Specifications

TurboGFP
Polyclonal
GFP2; Green Fluoresecent Protein; GFP; Turbo GFP; eGFP
Full-length recombit denatured TurboGFP and non-denatured CopGFP comprising 6XHis Tag
100 μg
Primary
Antibody
IgG
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Antigen affinity chromatography
RUO
-20°C
Lyophilized
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers