missing translation for 'onlineSavingsMsg'
Få mere at vide

anti-TRADD Clone: 7G8, Invitrogen™

Artikelnummer. 15374046
Skift visning
Klik for at se tilgængelige muligheder
Konjugeret:
Unconjugated
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Konjugeret unitSize
15374046 Unconjugated 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15374046 Leverandør Invitrogen Leverandørnr. A16595

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

TRADD Monoclonal antibody A16595 from Thermo Scientific™ Pierce™ specifically detects TRADD in Human samples, and it is validated for WB and IP

The protein encoded by this gene is a death domain containing adaptor molecule that interacts with TNFRSF1A/TNFR1 and mediates programmed cell death signaling and NF-kappaB activation. This protein binds adaptor protein TRAF2, reduces the recruitment of inhibitor-of-apoptosis proteins (IAPs) by TRAF2, and thus suppresses TRAF2 mediated apoptosis. This protein can also interact with receptor TNFRSF6/FAS and adaptor protein FADD/MORT1, and is involved in the Fas-induced cell death pathway.

A16595 detects TRADD in Human samples. A16595 has been successfully used in WB and IP procedures. The immunogen for A16595 is synthetic peptide corresponding to residues surrounding glycine 227 of human TRADD.

Applications:

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen TRADD
Klassifikation Monoclonal
Klon 7G8
Konjugeret Unconjugated
Genadgangsnr. Q15628
Værtsarter Rabbit
Immunogen synthetic peptide corresponding to residues surrounding glycine 227 of human TRADD.
Primær eller sekundær Primary
Gen-id (Entrez) 8717
Målarter Human
Indhold og opbevaring Store at -20°C
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.