missing translation for 'onlineSavingsMsg'
Learn More
Learn More
timeless homolog (Drosophila), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TIMELESS protein.
Brand: Abnova H00008914-B01P.50ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MDLHMMNCELLATCSALGYLEGDTYHKEPDCLESVKDLIRYLRHEDETRDVRQQLGAAQILQSDLLPILTQHHQDKPLFDAVIRLMVNLTQPALLCFGNLPKEPSFRHHFLQVLTYLQAYKEAFASEKAFGVLSETLYELLQLGWEERQEEDNLLIERILLLVRNILHVPADLDQEKKIDDDASAHDQLLWAIHLSGLDDLLLFLASSSAEEQWSLHVLEIVSLMFRDQNPEQLAGVGQGRLAQERSADFAELEVLRQREMAEKKTRALQRGNRHSRFGGSYIVQGLKSIGERDLIFHKGLHNLRNYSSDLGKQPKKVPKRRQAARELSIQRRSALNVRLFLRDFCSEFLENCYNRLMGSVKDHLLREKAQQHDETYYMWALAFFMAFNRAASFRPGLVSETLSVRTFHFIEQNLTNYYEMMLTDRKEAASWARRMHLALKAYQELLATVNEMDLSPDEAVRESSRIIKNNIFYVMEYRELFLALFRKFDERCQPRSFLRDLVETTHLFLKMLERFCRSRGNLVVQNKQKKRRKKKKKVLDQAIVSGNVPSSPEEVEAVWPALAEQLQCCAQNSELSMDSVVPFDAASEVPVEEQRAEAMVRIQDCLLAGQAPQALTLLRSAREVWPEGDVFGSQDISPEEEIQLLKQILSAPLPRQQGPEERGAEEEEEEEEEEEEELQVVQVSEKEFNFLDYLKRFACSTVVRAYVLLLRSYQQNSAHTNHCIVKMLHRLAHDLKMEALLFQLSVFCLFNRLLSDPAAGAYKELVTFAKYILGKFFALAAVNQKAFVELLFWKNTAVVREMTEGYGSLDDRSSSRRAPTWSPEEEAHLQELYLANKDVEGQDVVEAILAHLNTVPRTRKQIIHHLVQMGLADSVKDFQRKGTHIVLWTGDQELELQRLFEEFRDSDDVLGHIMKNITAKRSRARIVDKLLALGLVAERRELYKKRQKKLASSILPNGAESLKDFCQEDLEEEENLPEEDSEEEEEGGSEAEQVQGSLVLSNENLGQSLHQEGFSIPLLWLQNCLIRAADDREEDGCSQAVPLVPLTEENEEAMENEQFQQLLRKLGVRPPASGQETFWRIPAKLSPTQLRRAAASLSQPEEEQKLQPELQPKVPGEQGSDEEHCKEHRAQALRALLLAHKKKAGLASPEEEDAVGKEPLKAAPKKRQLLDSDEEQEEDEGRNRAPELGAPGIQKKKRYQIEDDEDDSpecifications
timeless homolog (Drosophila) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TIMELESS protein. | |
TIMELESS | |
FLJ12640/FLJ20714/TIM/TIM1/hTIM | |
Mouse | |
Affinity Purified | |
RUO | |
8914 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Immunofluorescence, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_003920.2 | |
TIMELESS | |
TIMELESS (NP_003911.1, 1 a.a. ∼ 1208 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- TIMELESS purified MaxPab mouse polyclonal antibody (B01P)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic