Få mere at vide
anti-TCEA3 Matched Antibody Pair, Abnova™
Beskrivelse
Indeholder
Capture antibody: mouse monoclonal anti-TCEA3 (100μg); Detection antibody: rabbit purified polyclonal anti-TCEA3 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TCEA3.
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Tekniske data
Tekniske data
| Antigen | TCEA3 |
| Applikationer | ELISA |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TCEA3. |
| Gene | TCEA3 |
| Gene Alias | TFIIS/TFIIS.H |
| Gensymboler | TCEA3 |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |