missing translation for 'onlineSavingsMsg'
Få mere at vide

T, Human, Matched Antibody Pair (AP51), Abnova™

Artikelnummer. 16162694
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16162694 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16162694 Leverandør Abnova Leverandørnr. H00006862AP51.1Set

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Indeholder

Capture antibody: mouse monoclonal anti-T (100μg); Detection antibody: rabbit purified polyclonal anti-T (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human T

Host Species: Mouse, Rabbit
Species Reactivity: Hu

ELISA Pair (Transfected Lysate)

Tekniske data

Antigen T
Applikationer ELISA
Klassifikation Monoclonal/Polyclonal
Konjugeret Unconjugated
Oversigt This antibody pair set comes with matched antibody pair to detect and quantify protein level of human T.
Gene T
Gene Alias MGC104817/TFT
Gensymboler T
Værtsarter Mouse, Rabbit
Mængde 1 Set
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 6862
Inkluderer Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-T (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-T (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Målarter Human
Indhold og opbevaring Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Purified
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.