Learn More
Cole-Parmer™ Anti-Static Dusting Brushes
Use this brush for a variety of brushing applications.
Brand: Cole-Parmer™ 46300-21
This item is not returnable.
View return policy
Description
- Anti-Static Dusting Brush
- 1 ÂĽ in. wide
Specifications
Anti-static Dusting Brush | |
Elimination of dust | |
1 Ea. | |
64 mm |
Product Content Correction
Product Title
Cole-Parmer™ Anti-Static Dusting Brushes > Bristle Width: 64mm
Spot an opportunity for improvement?Share a Content Correction