Learn More
Cole-Parmer™ Anti-Static Dusting Brushes
Use this brush for a variety of brushing applications.
Brand:  Cole-Parmer™ 46300-01
 View more versions of this product
Product Code. 11707603
  • 311.00 DKK / Each
Only null left
Estimated Shipment: 17-09-2025
Description
  • Anti-Static Dusting Brush
  • 1 ÂĽ in. wide
Specifications
Specifications
Anti-static Dusting Brush
Elimination of dust
1 Ea.
32 mm
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ Anti-Static Dusting Brushes > Bristle Width: 32mm