missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA4 Monoclonal Antibody (MC-813-70), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1021PE
Additional Details : Weight : 0.01000kg
Description
MA1-021-PE has been successfully used in ICC/IF and flow cytometry applications on human samples.
SSEA4 Monoclonal antibody specifically detects SSEA4 in Human samples. It is validated for Flow Cytometry.Specifications
SSEA4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG3 |
Flow Cytometry | |
MC-813-70 | |
PE | |
SSEA-4; stage-specific embryonic antigen 4 | |
human embryonal carcinoma cell line 2102Ep | |
100 μL | |
Primary | |
4° C, do not freeze | |
Liquid |