missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ SERPINA3G Monoclonal Antibody (MoFo 29.2)
GREENER_CHOICE

Artikelnummer. 11588762
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
Pakningsstørrelse:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11588762 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11588762 Leverandør Invitrogen™ Leverandørnr. MA1030

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Armenian Hamster Monoclonal Antibody

MA1-030 detects Spi2A in human, mouse and hamster samples. MA1-030 has been used successfully in Western blot, ICC/IF and ELISA procedures. By Western blot this antibody detects a ~49 kDa band representing the Spi2A protein. The MA1-030 immunogen is a synthetic peptide corresponding to residues N(418) P E R S M N F P N G E G A S S Q R L C(436) of hamster Spi2A protein.

Serine protease inhibitor 2A, or Serpin 2A (spi2A), inhibits lysosomal executioner proteases, thus enhancing the survival of cytotoxic T lymphocytes and allowing them to develop into memory CD8 T cells. Through its function of preserving T cells, spi2A has become a research target for such topics as immunity, tumor cell, and AIDS research.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen SERPINA3G
Applikationer ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon MoFo 29.2
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Serpina3g
Genadgangsnr. Q5I2A0
Gene Alias 2A2; AI119734; alpha-1 antiproteinase; serine (or cysteine) peptidase inhibitor, clade A, member 3G; serine (or cysteine) proteinase inhibitor, clade A, member 3G; serine protease inhibitor 2-1; serine protease inhibitor 2A; Serine protease inhibitor A3G; serpin 2A; Serpin A3G; Serpina3g; spi2; Spi2/eb.1; Spi2-1; Spi2A
Gensymboler Serpina3g
Værtsarter Armenian Hamster
Immunogen Synthetic peptide corresponding to residues N(418) P E R S M N F P N G E G A S S Q R L C(436) of hamster Spi2A protein.
Oprensningsmetode Protein A
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 20715
Målarter Human, Mouse, Hamster
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG2b
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.