Få mere at vide
anti-SMARCB1, BAZ1B Protein Protein Interaction Antibody Pair, Abnova™
Artikelnummer. 16062478
Skift visning
Mængde:
1 Set
Pakningsstørrelse:
1 sæt
Artikelnummer. Mængde unitSize
16062478 1 Set 1 sæt
1 options
Artikelnummer. 16062478 Leverandør Abnova Leverandørnr. DI0189.1Set
Indeholder
SMARCB1 rabbit purified polyclonal antibody (20μg); BAZ1B mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the SMARCB1 protein, and the other against the BAZ1B protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Tekniske data
Antigen SMARCB1
Applikationer In situ PLA
Klassifikation Monoclonal/Polyclonal
Konjugeret Unconjugated
Gene SMARCB1
Gene Alias BAF47/INI1/RDT/SNF5/SNF5L1/Sfh1p/Snr1/hSNFS
Gensymboler SMARCB1
Værtsarter Mouse, Rabbit
Mængde 1 Set
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 6598/9031
Inkluderer Antibody pair set content: 1. SMARCB1 rabbit purified polyclonal antibody (20 ug) 2. BAZ1B mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Målarter Human
Indhold og opbevaring Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Produkttype SMARCB1 & BAZ1B Protein Protein Interaction Antibody Pair
Form Purified