missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sucrase-isomaltase (alpha-glucosidase), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SI.
Brand: Abnova H00006476-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: DGESIDTYERDLYLSVQFNLNQTTLTSTILKRGYINKSETRLGSLHVWGKGTTPVNAVTLTYNGNKNSLPFNEDTTNMILRIDLTTHNVTLEEPIEINWSpecifications
sucrase-isomaltase (alpha-glucosidase) | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SI. | |
SI | |
MGC131621/MGC131622 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_001041 | |
SI | |
SI (NP_001032, 1728 a.a. ∼ 1826 a.a) partial recombinant protein with GST tag. | |
RUO | |
6476 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |