missing translation for 'onlineSavingsMsg'
Få mere at vide

SHC Mouse, Unlabeled, Clone: 30, BD

Artikelnummer. 15835689
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15835689 150 μg
15825689 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15835689 Leverandør BD Biosciences Leverandørnr. 610879

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

The mammalian SHC proteins, which are expressed as multiple isoforms (46, 52, and 66kDa), each contain a C-terminal SH2 domain and an N-terminal glycine/proline rich sequence. These proteins function as early signaling intermediates (also called adaptor proteins) which relay G protein coupled receptor (GPCR) and receptor tyrosine kinase (RTK)-induced signals via the Ras transduction pathway. To this end, the SHC proteins contain specific tyrosine residues which are phosphorylated following association with the active RTKs.
Phosphorylated SHC forms a complex with the adaptor protein GRB2. Association of the SHC-GRB2 complex with the Ras guanine nucleotide exchange factor (Ras-GEF) mediates the localization of Ras-GEF to the plasma membrane. Once at the plasma membrane, Ras-GEF activates Ras by catalyzing the Ras-GTP for Ras-GDP exchange. Over-expression of SHC results in cell transformation, and phosphorylation of SHC correlates with activation of the ERK1/ERK2 kinases. The SHC proteins are mediators of signals that are essential
for cell metabolism, growth, and differentiation.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen SHC
Applikationer Western Blot
Klassifikation Monoclonal
Klon 30
Koncentration 250μg/mL
Konjugeret Unconjugated
Formulering Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Værtsarter Mouse
Immunogen Human SHC aa. 359-473
Oprensningsmetode Affinity Purified
Mængde 150 μg
Regulatorisk status RUO
Forskningsdisciplin Cell Biology
Primær eller sekundær Primary
Målarter Canine, Chicken, Human, Mouse, Rat
Indhold og opbevaring Store undiluted at -20°C.
Isotype IgG1
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.