Få mere at vide
SGCG, Human, IP-WB Antibody Pair (PW3), Abnova™
Beskrivelse
Indeholder
Antibody pair for IP: rabbit polyclonal anti-SGCG (300μL), Antibody pair for WB: mouse purified polyclonal anti-SGCG (50μg)
Tekniske data
Tekniske data
| Antigen | SGCG |
| Applikationer | Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | SGCG |
| Gene Alias | A4/DAGA4/DMDA/DMDA1/LGMD2C/MAM/MGC130048/SCARMD2/SCG3/TYPE |
| Gensymboler | SGCG |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |
For Research Use Only