missing translation for 'onlineSavingsMsg'
Få mere at vide

serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Artikelnummer. 16172335
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16172335 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16172335 Leverandør Abnova Leverandørnr. H00005054D01P.100ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against a full-length human SERPINE1 protein.

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MQMSPALTCLVLGLALVFGEGSAVHHPPSYVAHLASDFGVRVFQQVAQASKDRNVVFSPYGVASVLAMLQLTTGGETQQQIQAAMGFKIDDKGMAPALRHLYKELMGPWNKDEISTTDAIFVQRDLKLVQGFMPHFFRLFRSTVKQVDFSEVERARFIINDWVKTHTKGMISNLLGKGAVDQLTRLVLVNALYFNGQWKTPFPDSSTHRRLFHKSDGSTVSVPMMAQTNKFNYTEFTTPDGHYYDILELPYHGDTLSMFIAAPYEKEVPLSALTNILSAQLISHWKGNMTRLPRLLVLPKFSLETEVDLRKPLENLGMTDMFRQFQADFTSLSDQEPLHVAQALQKVKIEVNESGTVASSSTAVIVSARMAPEEIIMDRPFLFVVRHNPTGTVLFMGQVMEP

Tekniske data

Antigen serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1
Applikationer Western Blot
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against a full-length human SERPINE1 protein.
Formulering PBS with no preservative; pH 7.4
Gene SERPINE1
Genadgangsnr. NM_000602
Gene Alias PAI/PAI-1/PAI1/PLANH1
Gensymboler SERPINE1
Værtsarter Rabbit
Immunogen SERPINE1 (NP_000593.1, 1 a.a. ∼ 402 a.a) full-length human protein.
Oprensningsmetode Affinity Purified
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 5054
Målarter Human
Indhold og opbevaring Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Produkttype Antibody
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.