missing translation for 'onlineSavingsMsg'
Learn More
Learn More
regulating synaptic membrane exocytosis 2, Mouse, Clone: 3C12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RIMS2.
Brand: Abnova H00009699-M01A.200uL
Additional Details : Weight : 0.00010kg
Description
Sequence: LANADLEDSRISMLRMDRPSRQRSISERRAAMENQRSYSMERTREAQGPSSYAQRTTNHSPPTPRRSPLPIDRPDLRRTDSpecifications
regulating synaptic membrane exocytosis 2 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_014677 | |
RIMS2 | |
RIMS2 (NP_055492, 181 a.a. ∼ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
Primary | |
Human | |
Antibody | |
IgM κ |
ELISA, Western Blot | |
3C12 | |
Mouse monoclonal antibody raised against a partial recombinant RIMS2. | |
RIMS2 | |
DKFZp781A0653/KIAA0751/OBOE/RAB3IP3/RIM2 | |
Mouse | |
200 μL | |
Yes | |
9699 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |