Få mere at vide
anti-RBM5, Human, IP-WB Antibody Pair, Abnova™
Beskrivelse
Indeholder
Antibody pair for IP: rabbit polyclonal anti-RBM5 (300μL), Antibody pair for WB: mouse purified polyclonal anti-RBM5 (50μg)
Tekniske data
Tekniske data
| Antigen | RBM5 |
| Applikationer | Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | RBM5 |
| Gene Alias | FLJ39876/G15/H37/LUCA15/RMB5 |
| Gensymboler | RBM5 |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |
For Research Use Only