missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beskrivelse
IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, In vitro assay.
Tekniske data
Tekniske data
| Antigen | IgG |
| Applikationer | ChIP Assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Klassifikation | Polyclonal |
| Koncentration | 0.5 mg/mL |
| Konjugeret | Unconjugated |
| Fortynding | Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 23034403), Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 32784470), Immunohistochemistry-Paraffin 1:10 - 1:500. Use reported in scientific literature (PMID 255994242), In vitro assay, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Formulering | PBS with 0.05% Sodium Azide |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gensymboler | IGHG1 |
| Værtsarter | Rabbit |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?