missing translation for 'onlineSavingsMsg'
Få mere at vide

Rab5 Mouse, Unlabeled, Clone: 15, BD

Artikelnummer. 15885379
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15885379 150 μg
15875379 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15885379 Leverandør BD Biosciences Leverandørnr. 610282

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Rab5 is a low molecular weight GTP-binding protein that plays a role in endocytic vesicle traffic. Like other Rab proteins, Rab5 has C-terminal cysteine residues that are post-translationally modified by geranylgeranylation, which is critical for its membrane targeting. Rab5 is associated with early endosome and plasma membranes and evidence suggests that Rab5 is involved in regulation of early endosome fusion. The GTP/GDP cycle controls shuttling of Rab proteins between the cytosol and organelle membranes. In vitro, Rab5 proteins are removed from membranes by a GDP dissociation inhibitor protein (rabGDI) which leads to the formation of a cytosolic Rab5-rabGDI complex. Rab5 insertion into membranes is a multistep process in which a transient GDP-Rab5 intermediate is formed and converted into GTP-Rab5 that subsequently enters the acceptor membrane and releases rabGDI into the cytosol.

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Rab5
Applikationer Western Blot
Klassifikation Monoclonal
Klon 15
Koncentration 250μg/mL
Konjugeret Unconjugated
Formulering Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Værtsarter Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Oprensningsmetode Affinity Purified
Mængde 150 μg
Regulatorisk status RUO
Forskningsdisciplin Cell Biology
Primær eller sekundær Primary
Målarter Human, Mouse, Rat
Indhold og opbevaring Store undiluted at -20°C.
Isotype IgG1
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.