Learn More
peroxisomal membrane protein 3, 35kDa, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PXMP3.
Brand: Abnova H00005828-A01.50uL
Additional Details : Weight : 0.00010kg
Description
This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq
Sequence: KAKLSSWCIPLTGAPNSDNTLATSGKECALCGEWPTMPHTIGCEHIFCYFCAKSSFLFDVYFTCPKCGTEVHSLQPLKSGIEMSEVNALSpecifications
peroxisomal membrane protein 3, 35kDa | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant PXMP3. | |
PXMP3 | |
PAF-1/PAF1/PEX2/PMP3/PMP35/RNF72 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human, Mouse | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000318 | |
PXMP3 | |
PXMP3 (NP_000309, 217 a.a. ∼ 305 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
5828 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |