missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Prokineticin Receptor 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534108
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Horse (95%) Elephant, Panda, Bovine (89%) Marmoset, Dog, Opossum (84%).
GPR73A is a receptor for prokineticin 1. It is a member of the GPCR family and is coupled to the G(q) subclass of heteromeric G proteins. Activation of the receptor leads to mobilization of calcium, stimulation of phosphoinositide turnover and activation of p44/p42 mitogen-activated protein kinase. GPR73A expression has been reported in adrenal, peripheral blood leukocytes, brain, colon, pancreas, prostate, rectum, salivary gland, small intestine, spleen, stomach, testis, and thyroid. No human ESTs have been identified.
Tekniske data
Prokineticin Receptor 1 | |
Polyclonal | |
Unconjugated | |
PROKR1 | |
EG-VEGF receptor; EG-VEGFR1; G protein-coupled receptor 73; G protein-coupled receptor ZAQ; GPCR73A; GPR73; GPR73a; G-protein coupled receptor 73; G-protein coupled receptor ZAQ; PKR1; PK-R1; Prokineticin receptor 1; Prokr1; ZAQ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10887 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8TCW9 | |
PROKR1 | |
Synthetic 19 amino acid peptide from C-terminus of human PROKR1. | |
50 μg | |
Primary | |
Human, Monkey | |
Antibody | |
IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Invitrogen™ Prokineticin Receptor 1 Polyclonal Antibody > 50 μg; Unconjugated
Ser du en mulighed for forbedring?Del en indholdskorrektion