Få mere at vide
anti-PLSCR3, Human, IP-WB Antibody Pair, Abnova™
Beskrivelse
Indeholder
Antibody pair for IP: rabbit polyclonal anti-PLSCR3 (300μL), Antibody pair for WB: mouse polyclonal anti-PLSCR3 (50μL)
Tekniske data
Tekniske data
| Antigen | PLSCR3 |
| Applikationer | Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | PLSCR3 |
| Gensymboler | PLSCR3 |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Regulatorisk status | RUO |
| Vis mere |
For Research Use Only