missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Phosphotyrosine Mouse, HRP, Clone: PY20, BD
Beskrivelse
Phosphorylation of specific tyrosine residues is the result of of activation or stimulation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be autophosphorylated kinases or certain cellular protein substrates that are regulated in oncogenesis or cell growth. Antibodies to phosphotyrosine provide one of the best tools for the detection and characterization of phosphotyrosine proteins.
Tekniske data
Tekniske data
| Antigen | Phosphotyrosine |
| Applikationer | Western Blot |
| Klassifikation | Monoclonal |
| Klon | PY20 |
| Koncentration | 250μg/mL |
| Konjugeret | HRP |
| Formulering | Aqueous buffered solution containing BSA and glycerol. |
| Værtsarter | Mouse |
| Oprensningsmetode | Affinity Purified |
| Mængde | 150 μg |
| Vis mere |
For Research Use Only.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?