Få mere at vide
anti-PBX2, Human, IP-WB Antibody Pair, Abnova™
Beskrivelse
Indeholder
Antibody pair for IP: rabbit polyclonal anti-PBX2 (300μL), Antibody pair for WB: mouse monoclonal anti-PBX2 (50μg)
Tekniske data
Tekniske data
| Antigen | PBX2 |
| Applikationer | Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal/Polyclonal |
| Konjugeret | Unconjugated |
| Oversigt | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | PBX2 |
| Gene Alias | G17/HOX12/PBX2MHC |
| Gensymboler | PBX2 |
| Værtsarter | Mouse, Rabbit |
| Mængde | 1 Set |
| Vis mere |
For Research Use Only