Learn More
Oxytocin Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Brand: Invitrogen PA118416
Additional Details : Weight : 0.01000kg
Description
Reconstitute in 50 μL sterile water. Centrifuge to remove any insoluble material. It is recommended that a thawed sample is stored at 4 °C for no longer than 2 weeks. Allocation of appropriate anti-bacterial agent can increase shelf life by several weeks. Diluted serum should be prepared as required. Long term stability requires storage, preferably in small aliquots at -20 °C or lower. Glycerol (1:1) can be added to neat serum for additional stability if intended use does not prevent this.
Oxytocin Polyclonal antibody specifically detects Oxytocin in Human, Ovine samples. It is validated for Immunohistochemistry (Frozen).Specifications
Oxytocin | |
Polyclonal | |
Unconjugated | |
OXT | |
neurophysin 1; neurophysin I; Ocytocin; Ot; OT-NPI; oxcytocin; OXT; OXT-NPI; Oxy; Oxytocin; oxytocin, prepro- (neurophysin I); oxytocin, prepropeptide; oxytocin/neurophysin (Oxt) gene, complete gene, complete cds; oxytocin/neurophysin 1 prepropeptide; oxytocin/neurophysin I prepropeptide; oxytocin-neurophysin 1; oxytocin-neurophysin I; oxytocin-neurophysin I, preproprotein; prepropeptide for oxytocin and neurophysin; unnamed protein product | |
Guinea Pig | |
50 μL | |
Primary | |
Human, Ovine | |
Antibody | |
IgG |
Immunohistochemistry (Frozen) | |
Conc. Not Determined | |
whole serum with no preservative | |
P01178, P13389 | |
OXT | |
Synthetic peptide corresponding to the amino acids 20 - 28 from human Oxytocin-neurophysin 1, conjugated to carrier protein. | |
RUO | |
443390, 5020 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |