missing translation for 'onlineSavingsMsg'
Få mere at vide

nNOS Mouse, Unlabeled, Clone: 16, BD

Artikelnummer. 15895389
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15895389 150 μg
15885389 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15895389 Leverandør BD Biosciences Leverandørnr. 610309

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+ levels and enhance calmodulin binding. Neuronal NOS (nNOS or bNOS) and endothelial NOS (ECNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin and are regulated in a similar manner. However, both have been shown to be distinct gene products of about 155kDa and 140kDa, respectively, and the human forms show 52% amino acid identity. Neuronal NOS and induced macrophage NOS (iNOS) share 51% amino acid homology with the greatest degree of divergence in the calmodulin binding domain. Neuronal NOS, a cytosolic protein present mainly in neural tissues, has been purified and characterized from rat cerebellum. The NO synthesized by this enzyme acts as a neurotransmitter. ECNOS has been cloned from human vascular endothelium as well as from bovine aortic endothelial cells (BAEC) and has a unique N-myristylation consensus sequence that may explain its membrane localization.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen nNOS
Applikationer Western Blot
Klassifikation Monoclonal
Klon 16
Koncentration 250μg/mL
Konjugeret Unconjugated
Oversigt NOS Type I; Neuronal Nitric Oxide Synthase
Formulering Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Værtsarter Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Oprensningsmetode Affinity Purified
Mængde 150 μg
Regulatorisk status RUO
Forskningsdisciplin Cell Biology
Primær eller sekundær Primary
Målarter Canine, Human, Mouse, Rat
Indhold og opbevaring Store undiluted at -20°C.
Isotype IgG2a
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.