Få mere at vide
nNOS Mouse, Unlabeled, Clone: 16, BD
Beskrivelse
Nitric oxide synthase (NOS), a cell-type specific enzyme, catalyzes the synthesis of nitric oxide (NO). NO is a short-lived radical that transmits cellular signals involved in vasorelaxation, neurotransmission, and cytotoxicity. In neurons and endothelial cells, constitutive NOS (cNOS) is activated by agonists that increase intracellular Ca2+ levels and enhance calmodulin binding. Neuronal NOS (nNOS or bNOS) and endothelial NOS (ECNOS) have recognition sites for NADPH, FAD, FMN, and calmodulin and are regulated in a similar manner. However, both have been shown to be distinct gene products of about 155kDa and 140kDa, respectively, and the human forms show 52% amino acid identity. Neuronal NOS and induced macrophage NOS (iNOS) share 51% amino acid homology with the greatest degree of divergence in the calmodulin binding domain. Neuronal NOS, a cytosolic protein present mainly in neural tissues, has been purified and characterized from rat cerebellum. The NO synthesized by this enzyme acts as a neurotransmitter. ECNOS has been cloned from human vascular endothelium as well as from bovine aortic endothelial cells (BAEC) and has a unique N-myristylation consensus sequence that may explain its membrane localization.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Tekniske data
Tekniske data
| Antigen | nNOS |
| Applikationer | Western Blot |
| Klassifikation | Monoclonal |
| Klon | 16 |
| Koncentration | 250μg/mL |
| Konjugeret | Unconjugated |
| Oversigt | NOS Type I; Neuronal Nitric Oxide Synthase |
| Formulering | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Værtsarter | Mouse |
| Immunogen | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Vis mere |
For Research Use Only.
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.