Learn More
NAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522298
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Rabbit (81%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NAT1 Polyclonal antibody specifically detects NAT1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
NAT1 | |
Polyclonal | |
Unconjugated | |
NAT1 | |
AAC1; Arylamide acetylase 1; arylamine N-acetyltransferase 1; AT-1; MNAT; Monomorphic arylamine N-acetyltransferase; N(alpha)-acetyltransferase 15, NatA auxiliary subunit; N-acetyl transferase 1; N-acetyl transferase 1, liver, blood; N-acetyltransferase (arylamine N-acetyltransferase); N-acetyltransferase 1; N-acetyltransferase 1 (arylamine N-acetyltransferase); N-acetyltransferase type 1; Nat; Nat1; NAT-1; NATI | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P18440 | |
NAT1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 187 of NAT1 (Uniprot ID#P18440) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |