Learn More
Myoglobin Monoclonal Antibody (6H8B5, 5A2G8, 4D7H3, 3H6A7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515305
Additional Details : Weight : 0.01000kg
Description
MA5-15305 targets Myoglobin in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15305 immunogen is purified recombinant fragment of Myoglobin expressed in E. Coli.
Myoglobin Monoclonal antibody specifically detects Myoglobin in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Myoglobin | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.03% sodium azide | |
P02144 | |
MB | |
Purified recombinant fragment of Myoglobin expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Western Blot | |
6H8B5, 5A2G8, 4D7H3, 3H6A7 | |
Unconjugated | |
MB | |
AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB | |
Mouse | |
Protein G | |
RUO | |
4151 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |