missing translation for 'onlineSavingsMsg'
Få mere at vide

MSH6 Mouse, Unlabeled, Clone: 44, BD

Artikelnummer. 15895699
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
15895699 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15895699 Leverandør BD Biosciences Leverandørnr. 610918

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

DNA mismatch repair in bacteria is carried out by the MutL, MutH, and MutS proteins. Initial binding of MutS to the mismatched DNA is followed by binding of the MutH endonuclease and MutL. Together these proteins form a complex that mediates excision repair. Mutations or deficiencies of any of these bacterial proteins results in a mutator phenotype that is characterized by genetic instability. MSH2, MSH3 and MSH6 are human homologs of MutS, while MLH1, PMS1, and PMS2 are homologs of MutL. As a heterodimer with MSH2, MSH6 binds to DNA containing G/T mismatches. The MSH2-MSH6 complex recognizes single-base mispairs and insertion/deletion loops. Binding of this complex induces conformational changes in the DNA that lead to the binding of an MLH-PMS1 complex and excision repair. Mutations in the human genes are associated with hereditary nonpolyposis colon cancer (HNPCC), a common hereditary disease in humans. HNPCC is characterized by frequent microsatellite mutations that arise from somatic mutation due to a replication error (RER+) phenotype. This phenotype is analogous to the bacterial system and is directly linked to DNA mismatch repair deficiencies.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen MSH6
Applikationer Western Blot
Klassifikation Monoclonal
Klon 44
Koncentration 250μg/mL
Konjugeret Unconjugated
Oversigt GTBP
Formulering Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Værtsarter Mouse
Immunogen Human MSH6 aa. 225-333
Oprensningsmetode Affinity Purified
Mængde 50 μg
Regulatorisk status RUO
Forskningsdisciplin Cell Biology
Primær eller sekundær Primary
Målarter Canine, Human, Mouse, Rat
Indhold og opbevaring Store undiluted at -20°C.
Isotype IgG1
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.