missing translation for 'onlineSavingsMsg'
Få mere at vide

mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Artikelnummer. 16180318
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16180318 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16180318 Leverandør Abnova Leverandørnr. H00004436D01P.100ug

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit polyclonal antibody raised against a full-length human MSH2 protein.

MSH2 was identified as a locus frequently mutated in hereditary nonpolyposis colon cancer (HNPCC). When cloned, it was discovered to be a human homolog of the E. coli mismatch repair gene mutS, consistent with the characteristic alterations in microsatellite sequences (RER+ phenotype) found in HNPCC. [provided by RefSeq]

Sequence: MAVQPKETLQLESAAEVGFVRFFQGMPEKPTTTVRLFDRGDFYTAHGEDALLAAREVFKTQGVIKYMGPAGAKNLQSVVLSKMNFESFVKDLLLVRQYRVEVYKNRAGNKASKENDWYLAYKASPGNLSQFEDILFGNNDMSASIGVVGVKMSAVDGQRQVGVGYVDSIQRKLGLCEFPDNDQFSNLEALLIQIGPKECVLPGGETAGDMGKLRQIIQRGGILITERKKADFSTKDIYQDLNRLLKGKKGEQMNSAVLPEMENQVAVSSLSAVIKFLELLSDDSNFGQFELTTFDFSQYMKLDIAAVRALNLFQGSVEDTTGSQSLAALLNKCKTPQGQRLVNQWIKQPLMDKNRIEERLNLVEAFVEDAELRQTLQEDLLRRFPDLNRLAKKFQRQAANLQDCYRLYQGINQLPNVIQALEKHEGKHQKLLLAVFVTPLTDLRSDFSKFQEMIETTLDMDQVENHEFLVKPSFDPNLSELREIMNDLEKKMQSTLISAARDLGLDPGKQIKLDSSAQFGYYFRVTCKEEKVLRNNKNFSTVDIQKNGVKFTNSKLTSLNEEYTKNKTEYEEAQDAIVKEIVNISSGYVEPMQTLNDVLAQLDAVVSFAHVSNGAPVPYVRPAILEKGQGRIILKASRHACVEVQDEIAFIPNDVYFEKDKQMFHIITGPNMGGKSTYIRQTGVIVLMAQIGCFVPCESAEVSIVDCILARVGAGDSQLKGVSTFMAEMLETASILRSATKDSLIIIDELGRGTSTYDGFGLAWAISEYIATKIGAFCMFATHFHELTALANQIPTVNNLHVTALTTEETLTMLYQVKKGVCDQSFGIHVAELANFPKHVIECAKQKALELEEFQYIGESQGYDIMEPAAKKCYLEREQGEKIIQEFLSKVKQMPFTEMSEENITIKLKQLKAEVIAKNNSFVNEIISRIKVTT

Tekniske data

Antigen mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli)
Applikationer Immunofluorescence, In situ PLA, Western Blot
Klassifikation Polyclonal
Konjugeret Unconjugated
Oversigt Rabbit polyclonal antibody raised against a full-length human MSH2 protein.
Formulering PBS with no preservative; pH 7.4
Gene MSH2
Genadgangsnr. NM_000251
Gene Alias COCA1/FCC1/HNPCC/HNPCC1/LCFS2
Gensymboler MSH2
Værtsarter Rabbit
Immunogen MSH2 (NP_000242.1, 1 a.a. ∼ 934 a.a) full-length human protein.
Oprensningsmetode Affinity Purified
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 4436
Målarter Human
Indhold og opbevaring Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Produkttype Antibody
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.