Learn More
membrane protein, palmitoylated 1, 55kDa, Mouse, Clone: 1E11-1G11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant MPP1.
Brand: Abnova H00004354-M01.100ug
Additional Details : Weight : 0.00010kg
Description
Palmitoylated membrane protein 1 is the prototype of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. Palmitoylated membrane protein 1 contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. [provided by RefSeq
Sequence: MTLKASEGESGGSMHTALSDLYLEHLLQKRSRPEAVSHPLNTVTEDMYTNGSPAPGSPAQVKGQEVRKVRLIQFEKVTEEPMGITLKLNEKQSCTVARILHGGMIHRQGSLHVGDEILEINGTNVTNHSVDQLQKAMKETKGMISLKVIPNQQSRLPALQMFMRAQFDYDPKKDNLIPCKEAGLKFATGDIIQIINKDDSNWWQGRVEGSSKESAGLIPSPELQEWRVASMAQSAPSEAPSCSPFGKKKKYKDKYLAKHSSIFDQLDVVSYEEVVRLPAFKRKTLVLIGASGVGRSHIKNALLSQNPEKFVYPVPYTTRPPRKSEEDGKEYHFISTEEMTRNISANEFLEFGSYQGNMFGTKFETVHQIHKQNKIAILDIEPQTLKIVRTAELSPFIVFIAPTDQGTQTEALQQLQKDSEAIRSQYAHYFDLSLVNNGVDETLKKLQEAFDQACSSPQWVPVSWVYSpecifications
membrane protein, palmitoylated 1, 55kDa | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC002392 | |
MPP1 | |
MPP1 (AAH02392, 1 a.a. ∼ 466 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
4354 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
1E11-1G11 | |
Mouse monoclonal antibody raised against a full length recombinant MPP1. | |
MPP1 | |
AAG12/DXS552/DXS552E/EMP55/MRG1/PEMP | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |