Learn More
Mouse IgG1 Isotype Control, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen PA533201
Additional Details : Weight : 0.01000kg
Description
PA5-33201 has been tested and found to be negative on the following rat tissues - peripheral blood leucocytes, thymocytes, splenocytes and macrophages. PA5-33201 recognizes the human CD45RA antigen, and therefore human leucocytes may be used as a positive control for this product. This antibody is not suitable for use as a negative control on human tissues. This product is a Low Endotoxin formulation. Endotoxin level is <0.01 EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Mouse IgG1 | |
1 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |
Control, Flow Cytometry | |
Unconjugated | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Human T lymphocytes | |
500 μg | |
Isotype Control (Primary) | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |